Anti-HEAB Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-15436R
BOSSBS-15436REA 725 EUR
BOSSBS-15436R
Anti-HEAB Rabbit Polyclonal Antibody
Antikörper
HEAB is a 425 amino acid nuclear protein that belongs to the Clp1 family. Utilizing magnesium, manganese or nickel as cofactors, HEAB participates in the phosphorylation of the 5'-hydroxyl groups of double- and single- stranded RNA and DNA. HEAB is a member of the tRNA splicing endonuclease complex, in conjunction with TSEN2, TSEN15, TSEN34 and TSEN54, and is also a member of the pre-mRNA cleavage complex II. The gene encoding HEAB maps to human chromosome 11q12.1 and mouse chromosome 2 D; mutations to this gene may lead to a reduced pre-mRNA cleavage activity. HEAB exists as two isoforms due to alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HEAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR