Anti-HEAB Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Lieferant: Bioss
BS-15436R-A350
BOSSBS-15436R-A350EA 893 EUR
BOSSBS-15436R-A350
Anti-HEAB Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antikörper
HEAB is a 425 amino acid nuclear protein that belongs to the Clp1 family. Utilising magnesium, manganese or nickel as cofactors, HEAB participates in the phosphorylation of the 5'-hydroxyl groups of double- and single- stranded RNA and DNA. HEAB is a member of the tRNA splicing endonuclease complex, in conjunction with TSEN2, TSEN15, TSEN34 and TSEN54, and is also a member of the pre-mRNA cleavage complex II. The gene encoding HEAB maps to human chromosome 11q12.1 and mouse chromosome 2 D; mutations to this gene may lead to a reduced pre-mRNA cleavage activity. HEAB exists as two isoforms due to alternative splicing events.
Type: Primary
Antigen: HEAB
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 1-100/425
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen