Anti-Hephaestin Rabbit Polyclonal Antibody (Cy7)

Lieferant: Bioss
BS-15458R-CY7
BOSSBS-15458R-CY7EA 883 EUR
BOSSBS-15458R-CY7
Anti-Hephaestin Rabbit Polyclonal Antibody (Cy7)
Antikörper
Hephaestin is a single-pass type I membrane protein that belongs to the multicopper oxidase family of proteins. Hephaestin, a copper-dependant ferroxidase protein, is crucial for iron exiting intestinal enterocytes into the circulation. It mediates the movement of iron across the basolateral membrane in conjunction with ferroportin 1. This is an important link between iron and copper metabolism in mammalian systems, as copper deficiency leads to reduced hephaestin and reduced iron absorption resulting in anemia. Hephaestin can bind six copper ions per monomer and is regulated by the homeobox transcription factor CDX2. Increased levels of iron leads to an increase in CDX2 expression and thus Hephaestin. Hephaestin is primarily detected in the intestine, but is also expressed in colon, breast, bone trabecural cells and fibroblasts.

Type: Primary
Antigen: Hephaestin
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 21-120/1158
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR