Anti-FOXO3A Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-1548R-A647
BOSSBS-1548R-A647EA 901 EUR
BOSSBS-1548R-A647
Anti-FOXO3A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Transcriptional activator which triggers apoptosis in the absence of survival factors, including neuronal cell death upon oxidative stress. recognises and binds to the DNA sequence 5'-[AG]TAAA[TC]A-3'. Participates in post-transcriptional regulation of MYC: following phosphorylation by MAPKAPK5, promotes induction of miR-34b and miR-34C expression, 2 post-transcriptional regulators of MYC that bind to the 3'UTR of MYC transcript and prevent its translation.

Type: Primary
Antigen: FOXO3A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 581-673/673
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR