Anti-LTK Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Lieferant: Bioss
BS-15500R-A488
BOSSBS-15500R-A488EA 893 EUR
BOSSBS-15500R-A488
Anti-LTK Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Antikörper
LTK is a member of the ros/insulin receptor family of tyrosine kinases. Tyrosine-specific phosphorylation of proteins is a key to the control of diverse pathways leading to cell growth and differentiation. Multiple transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: LTK
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 41-140/864
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR