Anti-TEC Rabbit Polyclonal Antibody (Cy7)

Lieferant: Bioss
BS-15505R-CY7
BOSSBS-15505R-CY7EA 900 EUR
BOSSBS-15505R-CY7
Anti-TEC Rabbit Polyclonal Antibody (Cy7)
Antikörper
TEC is a belongs to the Tec family of non-receptor protein-tyrosine kinases containing a pleckstrin homology domain. Tec family kinases are involved in the intracellular signaling mechanisms of cytokine receptors, lymphocyte surface antigens, heterotrimeric G-protein coupled receptors, and integrin molecules. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.

Type: Primary
Antigen: TEC
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 11-100/631
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR