Anti-TRP1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)

Lieferant: Bioss
BS-15510R-A680
BOSSBS-15510R-A680EA 874 EUR
BOSSBS-15510R-A680
Anti-TRP1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antikörper
Oxidation of 5,6-dihydroxyindole-2-carboxylic acid (DHICA) into indole-5,6-quinone-2-carboxylic acid. May regulate or influence the type of melanin synthesised.

Type: Primary
Antigen: TRP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/537
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR