Anti-E.coli O139 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1555R
BOSSBS-1555REA 1090 EUR
BOSSBS-1555R
Anti-E.coli O139 Rabbit Polyclonal Antibody
Antikörper
This antibody eacts with Escherichia coli O139 (E.coli O139), Escherichia coli is a gram negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: E.coli O139
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR