Anti-E.coli O139 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))

Lieferant: Bioss
BS-1555R-HRP
BOSSBS-1555R-HRPEA 1350 EUR
BOSSBS-1555R-HRP
Anti-E.coli O139 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Antikörper
This antibody eacts with Escherichia coli O139 (E.coli O139), Escherichia coli is a gram negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins.

Type: Primary
Antigen: E.coli O139
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR