Anti-IGFBPL1 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-15569R
BOSSBS-15569REA 725 EUR
BOSSBS-15569R
Anti-IGFBPL1 Rabbit Polyclonal Antibody
Antikörper
IGFBPL1 is a secreted IGF (Insulin-like growth factor) binding protein that is known to contain an Ig-like C2-type (immunoglobulin-like) domain, an IGFBP N-terminal domain and a Kazal-like domain. IGF-binding proteins characteristically act to extend the half-life of IGFs and may influence the growth promoting effects of the IGFs. The interaction of IGFBPs with IGFs can affect cell surface receptors, specifically, IGFBPs may enhance or decrease a cells insulin sensitivity. IGFBPL1 has been found to be down-regulated in multiple tumors and thus may be a likely tumor suppressor candidate. Highly expressed in both brain and testis, IGFBPL1 is found at lower levels in the prostate, bladder and lung.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: IGFBPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat