Anti-IkB beta Ser19 Rabbit Polyclonal Antibody (Cy3)

Lieferant: Bioss
BS-15581R-CY3
BOSSBS-15581R-CY3EA 900 EUR
BOSSBS-15581R-CY3
Anti-IkB beta Ser19 Rabbit Polyclonal Antibody (Cy3)
Antikörper
The protein encoded by this gene belongs to the NF-kappa-B inhibitor family, which inhibit NF-kappa-B by complexing with, and trapping it in the cytoplasm. Phosphorylation of serine residues on these proteins by kinases marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B, which translocates to the nucleus to function as a transcription factor. Alternatively spliced transcript variants have been found for this gene.

Type: Primary
Antigen: IkB beta Ser19
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR