Anti-GPAT4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)

Lieferant: Bioss
BS-15587R-A647
BOSSBS-15587R-A647EA 893 EUR
BOSSBS-15587R-A647
Anti-GPAT4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Antikörper
Lysophosphatidic acid acyltransferases (EC 2,3,1,51) catalyse the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.

Type: Primary
Antigen: GPAT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 251-350/648
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR