Anti-GPAT4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)

Lieferant: Bioss
BS-15587R-A680
BOSSBS-15587R-A680EA 890 EUR
BOSSBS-15587R-A680
Anti-GPAT4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antikörper
Lysophosphatidic acid acyltransferases (EC 2,3,1,51) catalyse the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.

Type: Primary
Antigen: GPAT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/648
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

TECHNISCHE DATEN

Environmentally Preferable
Antigen-Name GPAT4
Antigen-Synonyme LPAAT zeta|Glycerol-3-phosphate acyltransferase 4|GPAT4|Acyl-CoA:glycerol-3-phosphate acyltransferase 4|1-AGP acyltransferase 6|1-acylglycerol-3-phosphate O-acyltransferase 6|1-AGPAT 6|Lysophosphatidic acid acyltransferase zeta|LPAAT-zeta|GPAT4_HUMAN|1 AGP acyltransferase 6|1 AGPAT 6|AGPAT6|1 acyl sn glycerol 3 phosphate acyltransferase zeta
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 680
Reaktivität Mouse
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 251-350/648
Immunogen KLH conjugated synthetic peptide derived from human GPAT4
Form Liquid
Molecular weight 48 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR