Anti-GPAT4 Rabbit Polyclonal Antibody (Cy5)

Lieferant: Bioss
BS-15587R-CY5
BOSSBS-15587R-CY5EA 900 EUR
BOSSBS-15587R-CY5
Anti-GPAT4 Rabbit Polyclonal Antibody (Cy5)
Antikörper
Lysophosphatidic acid acyltransferases (EC 2,3,1,51) catalyse the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.

Type: Primary
Antigen: GPAT4
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 251-350/648
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR