Anti-LETM1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Lieferant: Bioss
BS-15591R-A488
BOSSBS-15591R-A488EA 893 EUR
BOSSBS-15591R-A488
Anti-LETM1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Antikörper
This gene encodes a protein that is localised to the inner mitochondrial membrane. The protein functions to maintain the mitochondrial tubular shapes and is required for normal mitochondrial morphology and cellular viability. Mutations in this gene cause Wolf-Hirschhorn syndrome, a complex malformation syndrome caused by the deletion of parts of the distal short arm of chromosome 4. Related pseudogenes have been identified on chromosomes 8, 15 and 19.

Type: Primary
Antigen: LETM1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 101-200/739
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR