Anti-E.coli O157:H7 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))

Lieferant: Bioss
BS-1563R-HRP
BOSSBS-1563R-HRPEA 883 EUR
BOSSBS-1563R-HRP
Anti-E.coli O157:H7 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Antikörper
This antibody reacts with Escherichia coli O157 (E.coli O157), Escherichia coli is a gram negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins.

Type: Primary
Antigen: E.coli O157:H7
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR