Anti-APOH Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-1570R-A647
BOSSBS-1570R-A647EA 893 EUR
BOSSBS-1570R-A647
Anti-APOH Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Binds to various kinds of negatively charged substances such as heparin, phospholipids, and dextran sulfate. May prevent activation of the intrinsic blood coagulation cascade by binding to phospholipids on the surface of damaged cells.

Type: Primary
Antigen: APOH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 281-345/345
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR