Anti-APOH Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1570R-CY3
BOSSBS-1570R-CY3EA 900 EUR
BOSSBS-1570R-CY3
Anti-APOH Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Binds to various kinds of negatively charged substances such as heparin, phospholipids and dextran sulfate. May prevent activation of the intrinsic blood coagulation cascade by binding to phospholipids on the surface of damaged cells.

Type: Primary
Antigen: APOH
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range:281-345/345
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR