Anti-factor 13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-1587R-A680
BOSSBS-1587R-A680EA 890 EUR
BOSSBS-1587R-A680
Anti-factor 13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Factor XIII is activated by thrombin and calcium ion to a transglutaminase that catalyses the formation of gamma-glutamyl-epsilon-lysine cross-links between fibrin chains, thus stabilizing the fibrin clot. Also cross-link alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin.
Type: Primary
Antigen: factor 13
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 331-732/732
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse