Anti-factor 13 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-1587R-CY5
BOSSBS-1587R-CY5EA 900 EUR
BOSSBS-1587R-CY5
Anti-factor 13 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Factor XIII is activated by thrombin and calcium ion to a transglutaminase that catalyses the formation of gamma-glutamyl-epsilon-lysine cross-links between fibrin chains, thus stabilizing the fibrin clot. Also cross-link alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin.

Type: Primary
Antigen: factor 13
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 331-732/732
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR