Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-1602R-CY5.5
BOSSBS-1602R-CY5.5EA 900 EUR
BOSSBS-1602R-CY5.5
Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localised to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterised to date.

Type: Primary
Antigen: SPA-1/SIPA-1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5
Public Immunogen Range: 351-450/1042
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR