Anti-Smad1 + Smad5 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-1619R
BOSSBS-1619REA 704 EUR
BOSSBS-1619R
Anti-Smad1 + Smad5 Rabbit Polyclonal Antibody
Antikörper
Transcriptional modulator activated by BMP (bone morphogenetic proteins) type 1 receptor kinase. SMAD5 is a receptor-regulated SMAD (R-SMAD). Transcriptional modulator activated by BMP (bone morphogenetic proteins) type 1 receptor kinase. SMAD1 is a receptor-regulated SMAD (R-SMAD). SMAD1/OAZ1/PSMB4 complex mediates the degradation of the CREBBP/EP300 repressor SNIP1. May act synergistically with SMAD4 and YY1 in bone morphogenetic protein (BMP)-mediated cardiac-specific gene expression.
Type: Primary
Antigen: Smad1 + Smad5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Public Immunogen Range: 361-465/465
Host: Rabbit
Isotype: IgG
Reactivity: Human