Anti-Smad1 + Smad5 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-1619R-CY5.5
BOSSBS-1619R-CY5.5EA 883 EUR
BOSSBS-1619R-CY5.5
Anti-Smad1 + Smad5 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Transcriptional modulator activated by BMP (bone morphogenetic proteins) type 1 receptor kinase. SMAD5 is a receptor-regulated SMAD (R-SMAD). Transcriptional modulator activated by BMP (bone morphogenetic proteins) type 1 receptor kinase. SMAD1 is a receptor-regulated SMAD (R-SMAD). SMAD1/OAZ1/PSMB4 complex mediates the degradation of the CREBBP/EP300 repressor SNIP1. May act synergistically with SMAD4 and YY1 in bone morphogenetic protein (BMP)-mediated cardiac-specific gene expression.

Type: Primary
Antigen: Smad1 + Smad5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5
Public Immunogen Range: 361-465/465
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR