Anti-B3GAT1/CD57 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-1635R-CY7
BOSSBS-1635R-CY7EA 909 EUR
BOSSBS-1635R-CY7
Anti-B3GAT1/CD57 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Involved in the biosynthesis of L2/HNK-1 carbohydrate epitope on glycoproteins. Can also play a role in glycosaminoglycan biosynthesis. Substrates include asialo-orosomucoid (ASOR), asialo-fetuin, and asialo-neural cell adhesion molecule. Requires sphingomyelin for activity: stearoyl-sphingomyelin was the most effective, followed by palmitoyl-sphingomyelin and lignoceroyl-sphingomyelin. Activity was demonstrated only for sphingomyelin with a saturated fatty acid and not for that with an unsaturated fatty acid, regardless of the length of the acyl group (By similarity).

Type: Primary
Antigen: B3GAT1/CD57
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 21-120/334
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR