Anti-IDO2 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)

Lieferant: Bioss
BS-16640R-A680
BOSSBS-16640R-A680EA 890 EUR
BOSSBS-16640R-A680
Anti-IDO2 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antikörper
Catalyses the first and rate-limiting step in the kynurenine pathway of tryptophan catabolism.

Type: Primary
Antigen: IDO2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/407
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR