Anti-IDO2 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Lieferant: Bioss
BS-16640R-A750
BOSSBS-16640R-A750EA 890 EUR
BOSSBS-16640R-A750
Anti-IDO2 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
Catalyses the first and rate-limiting step in the kynurenine pathway of tryptophan catabolism.
Type: Primary
Antigen: IDO2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/407
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse