Anti-XBP1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-1668R-CY5
BOSSBS-1668R-CY5EA 909 EUR
BOSSBS-1668R-CY5
Anti-XBP1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Transcription factor essential for hepatocyte growth, the differentiation of plasma cells, the immunoglobulin secretion, and the unfolded protein response (UPR). Acts during endoplasmic reticulum stress (ER) by activating unfolded protein response (UPR) target genes via direct binding to the UPR element (UPRE). Binds DNA preferably to the CRE-like element 5'-GATGACGTG[TG]N(3)[AT]T-3', and also to some TPA response elements (TRE). Binds to the HLA DR-alpha promoter. Binds to the Tax-responsive element (TRE) of HTLV-I.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: XBP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat