Anti-LIG4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-1677R-A350
BOSSBS-1677R-A350EA 893 EUR
BOSSBS-1677R-A350
Anti-LIG4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Efficiently joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. Involved in DNA non-homologous end joining (NHEJ) required for double-strand break repair and V(D)J recombination. The LIG4-XRCC4 complex is responsible for the NHEJ ligation step, and XRCC4 enhances the joining activity of LIG4. Binding of the LIG4-XRCC4 complex to DNA ends is dependent on the assembly of the DNA-dependent protein kinase complex DNA-PK to these DNA ends.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LIG4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat