Anti-SKI Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-1684R-A350
BOSSBS-1684R-A350EA 893 EUR
BOSSBS-1684R-A350
Anti-SKI Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
SKI is a nuclear protein that forms homodimers and heterodimers and binds to DNA to function as transcriptional activators and repressors. The SKI oncoprotein dramatically affects cell growth, differentiation, and/or survival. SKI was shown to act in distinct signaling pathways including those involving nuclear receptors, transforming growth factor beta, and tumor suppressors. SKI inhibits transforming growth factor-beta (TGF-beta) signaling through interaction with Smad proteins. SKI represses Smad-mediated transcriptional activation, probably through its action as a transcriptional co-repressor. SKI also inhibits TGF-beta-induced downregulation of genes such as c-myc.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SKI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR