Anti-SKI Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-1684R-CY5.5
BOSSBS-1684R-CY5.5EA 900 EUR
BOSSBS-1684R-CY5.5
Anti-SKI Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
SKI is a nuclear protein that forms homodimers and heterodimers and binds to DNA to function as transcriptional activators and repressors. The SKI oncoprotein dramatically affects cell growth, differentiation, and/or survival. SKI was shown to act in distinct signaling pathways including those involving nuclear receptors, transforming growth factor beta, and tumor suppressors. SKI inhibits transforming growth factor-beta (TGF-beta) signaling through interaction with Smad proteins. SKI represses Smad-mediated transcriptional activation, probably through its action as a transcriptional co-repressor. SKI also inhibits TGF-beta-induced downregulation of genes such as c-myc.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SKI
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat