Anti-SKI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-1684R-HRP
BOSSBS-1684R-HRPEA 900 EUR
BOSSBS-1684R-HRP
Anti-SKI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
SKI is a nuclear protein that forms homodimers and heterodimers and binds to DNA to function as transcriptional activators and repressors. The SKI oncoprotein dramatically affects cell growth, differentiation, and/or survival. SKI was shown to act in distinct signaling pathways including those involving nuclear receptors, transforming growth factor beta, and tumor suppressors. SKI inhibits transforming growth factor-beta (TGF-beta) signaling through interaction with Smad proteins. SKI represses Smad-mediated transcriptional activation, probably through its action as a transcriptional co-repressor. SKI also inhibits TGF-beta-induced downregulation of genes such as c-myc.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SKI
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR