Anti-PAI1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-1704R-CY5.5
BOSSBS-1704R-CY5.5EA 900 EUR
BOSSBS-1704R-CY5.5
Anti-PAI1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Serine protease inhibitor. This inhibitor acts as 'bait' for tissue plasminogen activator, urokinase, protein C and matriptase-3/TMPRSS7. Its rapid interaction with PLAT may function as a major control point in the regulation of fibrinolysis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAI1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR