Anti-DAT/Dopamine Transporter Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-1714R-A750
BOSSBS-1714R-A750EA 899 EUR
BOSSBS-1714R-A750
Anti-DAT/Dopamine Transporter Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Amine transporter. Terminates the action of dopamine by its high affinity sodium-dependent reuptake into presynaptic terminals.

Type: Primary
Antigen: DAT/Dopamine transporter
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 579-620/620
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR