Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1719R
BOSSBS-1719REA 725 EUR
BOSSBS-1719R
Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HPV16 E6 + HPV18 E6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR