Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-1719R-A350
BOSSBS-1719R-A350EA 893 EUR
BOSSBS-1719R-A350
Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Transcriptional transactivator. Binds double-stranded DNA (By similarity). Has transforming activity. Inactivates, with E6-AP ubiquitin-protein ligase, the human p53/TP53 tumor suppressor protein by targeting it to degradation. Binds and targets human MUPP1/MPDZ protein to degradation. Those two functions presumably contribute to transforming activity. Interaction with human FBLN1 protein also seems to be linked to cell transformation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPV16 E6 + HPV18 E6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR