Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1719R-A680
BOSSBS-1719R-A680EA 890 EUR
BOSSBS-1719R-A680
Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Human Papilloma virus (HPV) is responsible for the development of type and tissue specific papilloma in the oral cavity and in the larynx. In the skin papilloma virus can cause different types of warts and in the ano-genital region it is associated with condyloma or carcinoma. HPV types 16 and 18 seem to be responsible for the development of some carcinoma like cervical carcinoma.

Type: Primary
Antigen: HPV16 E6 + HPV18 E6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Virus, Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR