Anti-DUT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-1721R-A350
BOSSBS-1721R-A350EA 893 EUR
BOSSBS-1721R-A350
Anti-DUT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
This enzyme is involved in nucleotide metabolism: it produces dUMP, the immediate precursor of thymidine nucleotides and it decreases the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DUT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat