Anti-DUT Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-1721R-CY5
BOSSBS-1721R-CY5EA 900 EUR
BOSSBS-1721R-CY5
Anti-DUT Rabbit Polyclonal Antibody (Cy5®)
Antikörper
This enzyme is involved in nucleotide metabolism: it produces dUMP, the immediate precursor of thymidine nucleotides and it decreases the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUT
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR