Anti-CFH Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1745R
BOSSBS-1745REA 637 EUR
BOSSBS-1745R
Anti-CFH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CFH
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR