Anti-DRD4 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-1746R
BOSSBS-1746REA 725 EUR
BOSSBS-1746R
Anti-DRD4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DRD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat