Anti-DRD4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1746R-A680
BOSSBS-1746R-A680EA 890 EUR
BOSSBS-1746R-A680
Anti-DRD4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene encodes the D4 subtype of the dopamine receptor. The D4 subtype is a G-protein coupled receptor which inhibits adenylyl cyclase. It is a target for drugs which treat schizophrenia and Parkinson disease. Mutations in this gene have been associated with various behavioral phenotypes, including autonomic nervous system dysfunction, attention deficit/hyperactivity disorder, and the personality trait of novelty seeking. This gene contains a polymorphic number (2-10 copies) of tandem 48 nt repeats; the sequence shown contains four repeats.DRD4 expression has been reported in various regions of the brain as well as in adrenal gland, artery, eye, heart, kidney, placenta, spinal cord, testis, and vas deferens.

Type: Primary
Antigen: DRD4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/467
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR