Anti-OTUD5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Lieferant: Bioss
BS-17566R-A680
BOSSBS-17566R-A680EA 890 EUR
BOSSBS-17566R-A680
Anti-OTUD5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antikörper
Deubiquitinating enzyme that functions as negative regulator of the innate immune system. Acts via TRAF3 deubiquitination and subsequent suppression of type I interferon (IFN) production. Has peptidase activity towards 'Lys-48'- and 'Lys-63'-linked polyubiquitin chains. Can also cleave 'Lys-11'-linked ubiquitin chains (in vitro).
Type: Primary
Antigen: OTUD5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 501-571/571
Host: Rabbit
Isotype: IgG
Reactivity: Human