Anti-GATA4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-1778R-A350
BOSSBS-1778R-A350EA 876 EUR
BOSSBS-1778R-A350
Anti-GATA4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Transcriptional activator that binds to the consensus sequence 5'-AGATAG-3' and plays a key role in cardiac development. Involved in bone morphogenetic protein (BMP)-mediated induction of cardiac-specific gene expression. Binds to BMP response element (BMPRE) DNA sequences within cardiac activating regions. Acts as a transcriptional activator of ANF in cooperation with NKX2-5. Promotes cardiac myocyte enlargement. Required during testicular development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GATA4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR