Anti-GATA4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-1778R-A750
BOSSBS-1778R-A750EA 874 EUR
BOSSBS-1778R-A750
Anti-GATA4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Transcriptional activator that binds to the consensus sequence 5'-AGATAG-3' and plays a key role in cardiac development. Involved in bone morphogenetic protein (BMP)-mediated induction of cardiac-specific gene expression. Binds to BMP response element (BMPRE) DNA sequences within cardiac activating regions. Acts as a transcriptional activator of ANF in cooperation with NKX2-5. Promotes cardiac myocyte enlargement. Required during testicular development.
Type: Primary
Antigen: GATA4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 261-360/442
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat