Anti-GRIA3 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-1799R-CY7
BOSSBS-1799R-CY7EA 900 EUR
BOSSBS-1799R-CY7
Anti-GRIA3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for glutamate that functions as ligand-gated ion channel in the central nervous system and plays an important role in excitatory synaptic transmission. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter L-glutamate induces a conformation change, leading to the opening of the cation channel, and thereby converts the chemical signal to an electrical impulse. The receptor then desensitizes rapidly and enters a transient inactive state, characterized by the presence of bound agonist. In the presence of CACNG4 or CACNG7 or CACNG8, shows resensitization which is characterized by a delayed accumulation of current flux upon continued application of glutamate.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GLUR3/GRIA 3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat