Anti-P2RY4 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1857R
BOSSBS-1857REA 725 EUR
BOSSBS-1857R
Anti-P2RY4 Rabbit Polyclonal Antibody
Antikörper
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to uridine nucleotides, partially responsive to ATP, and not responsive to ADP. P2Y4 expression has been documented in blood, bone, heart, lung, pancreas, placenta, and umbilical cord.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: P2RY4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR