Anti-PDEF/SPDEF Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-1866R-A555
BOSSBS-1866R-A555EA 893 EUR
BOSSBS-1866R-A555
Anti-PDEF/SPDEF Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
SPDEF, also known as PDEF is an ETS transcription factor expressed in the prostate epithelial cells. It is thought to act as an androgen-independent transactivator of the prostate-specific antigen (PSA) promoter and also as a transcriptional activator for the SERPINB5 promoter. It is highly expressed in prostate carcinoma cells and is thought to be involved in the regulation of the prostate gland and/or prostate cancer development. It is also expressed in epithelial breast carcinoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDEF/SPDEF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR