Anti-GPAT4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-1924R-A555
BOSSBS-1924R-A555EA 893 EUR
BOSSBS-1924R-A555
Anti-GPAT4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
AGPAT6 is a novel glycerolipid acyltransferase of the ER, which is crucial for the production of milk fat by the mammary gland. It converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSARG7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR