Anti-ADH/AVP/ARVP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1930R-A680
BOSSBS-1930R-A680EA 874 EUR
BOSSBS-1930R-A680
Anti-ADH/AVP/ARVP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Vasopressin, also known as arginine vasopressin (AVP) or antidiuretic hormone (ADH), is a posterior pituitary hormone that is synthesised in the hypothalamus. Vasopressin is synthesised as a precursor protein that consists of arginine vasopressin and two associated proteins, neurophysin 2 and the glycopeptide copeptin. Vasopressin, together with its carrier protein neurophysin II, is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. Vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. It has a direct antidiuretic action on the kidney and also causes vasoconstriction of the peripheral vessels. Vasopressin can also contract smooth muscle during parturition and lactation. It also plays a role in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions. Mutations in the vasopressin precursor cause autosomal dominant neurohypophyseal diabetes insipidus (ADNDI), which is characterised by persistant thirst, polydipsia and polyuria.

Type: Primary
Antigen: ADH/AVP/ARVP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR