Anti-ZFP91 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BS-1941R-HRP
BOSSBS-1941R-HRPEA 900 EUR
BOSSBS-1941R-HRP
Anti-ZFP91 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The protein encoded by ZFP91 is a member of the zinc finger family of proteins. This protein contains C2H2 type domains, which are the classical zinc finger domains found in numerous nucleic acid-binding proteins. May be involved in transcriptional regulation. May play an important role in cell proliferation and/or anti-apoptosis Found in all the examined tissues including brain, heart, kidney, lung, liver, spleen, thymus, skeletal muscle, ovary and testis. Belongs to the krueppel C2H2-type zinc-finger protein family.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: ZFP91
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat